Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605143 MIM: 607035 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACTR1A - ARP1 actin-related protein 1 homolog A, centractin alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUFU - SUFU negative regulator of hedgehog signaling | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178133.1 | Intron | NP_001171604.1 | ||||
NM_016169.3 | Intron | NP_057253.2 | ||||
XM_011539858.2 | Intron | XP_011538160.1 | ||||
XM_011539860.2 | Intron | XP_011538162.1 | ||||
XM_011539861.2 | Intron | XP_011538163.1 | ||||
XM_011539863.2 | Intron | XP_011538165.1 | ||||
XM_011539864.2 | Intron | XP_011538166.1 | ||||
XM_017016323.1 | Intron | XP_016871812.1 |
Set Membership: |
HapMap Validated |