Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGGTTAGATGGGCAGCGGTTCC[A/G]TGGACAGAGGAGGAGGCGCGGCTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604430 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
GNG7 - G protein subunit gamma 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052847.2 | 4016 | UTR 3 | NP_443079.1 | |||
XM_017026606.1 | 4016 | UTR 3 | XP_016882095.1 |
LOC101929097 - uncharacterized LOC101929097 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005259695.2 | 4016 | UTR 3 | XP_005259752.1 |
Set Membership: |
HapMap |