Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
76 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605551 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf111 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf111 - chromosome 1 open reading frame 111 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182581.3 | 586 | Silent Mutation | AGA,AGG | R,R 174 | NP_872387.2 | |
XM_005245103.3 | 586 | Silent Mutation | AGA,AGG | R,R 136 | XP_005245160.1 | |
XM_011509443.2 | 586 | Silent Mutation | AGA,AGG | R,R 193 | XP_011507745.1 | |
XM_011509444.1 | 586 | Silent Mutation | AGA,AGG | R,R 124 | XP_011507746.1 | |
XM_011509445.2 | 586 | Intron | XP_011507747.1 |
C1orf226 - chromosome 1 open reading frame 226 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOS1AP - nitric oxide synthase 1 adaptor protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |