Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ULK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ULK4 - unc-51 like kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322500.1 | Intron | NP_001309429.1 | ||||
NM_001322501.1 | Intron | NP_001309430.1 | ||||
NM_017886.3 | Intron | NP_060356.2 | ||||
XM_011533874.2 | Intron | XP_011532176.1 | ||||
XM_011533880.2 | Intron | XP_011532182.1 | ||||
XM_017006711.1 | Intron | XP_016862200.1 | ||||
XM_017006712.1 | Intron | XP_016862201.1 |