Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610796 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPA4L PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HSPA4L - heat shock protein family A (Hsp70) member 4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317381.1 | Intron | NP_001304310.1 | ||||
NM_001317382.1 | Intron | NP_001304311.1 | ||||
NM_001317383.1 | Intron | NP_001304312.1 | ||||
NM_014278.3 | Intron | NP_055093.2 | ||||
XM_011531745.2 | Intron | XP_011530047.1 |
SLC25A31 - solute carrier family 25 member 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |