Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602887 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DLG4 - discs large MAGUK scaffold protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128827.2 | Intron | NP_001122299.1 | ||||
NM_001321074.1 | Intron | NP_001308003.1 | ||||
NM_001321075.1 | Intron | NP_001308004.1 | ||||
NM_001321076.1 | Intron | NP_001308005.1 | ||||
NM_001321077.1 | Intron | NP_001308006.1 | ||||
NM_001365.4 | Intron | NP_001356.1 | ||||
XM_005256491.1 | Intron | XP_005256548.1 | ||||
XM_011523698.1 | Intron | XP_011522000.1 | ||||
XM_011523699.2 | Intron | XP_011522001.1 | ||||
XM_011523702.1 | Intron | XP_011522004.1 | ||||
XM_017024288.1 | Intron | XP_016879777.1 | ||||
XM_017024289.1 | Intron | XP_016879778.1 | ||||
XM_017024290.1 | Intron | XP_016879779.1 |
Set Membership: |
HapMap |