Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 184420 MIM: 608933 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FDFT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FDFT1 - farnesyl-diphosphate farnesyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287742.1 | Intron | NP_001274671.1 | ||||
NM_001287743.1 | Intron | NP_001274672.1 | ||||
NM_001287744.1 | Intron | NP_001274673.1 | ||||
NM_001287745.1 | Intron | NP_001274674.1 | ||||
NM_001287747.1 | Intron | NP_001274676.1 | ||||
NM_001287748.1 | Intron | NP_001274677.1 | ||||
NM_001287749.1 | Intron | NP_001274678.1 | ||||
NM_001287750.1 | Intron | NP_001274679.1 | ||||
NM_001287751.1 | Intron | NP_001274680.1 | ||||
NM_001287756.1 | Intron | NP_001274685.1 | ||||
NM_004462.4 | Intron | NP_004453.3 | ||||
XM_017013217.1 | Intron | XP_016868706.1 |
NEIL2 - nei like DNA glycosylase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |