Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 616380 | |||||||||||||||||||||||||||||
Literature Links: |
LAMB4 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
LAMB4 - laminin subunit beta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318046.1 | 5298 | Intron | NP_001304975.1 | |||
NM_001318047.1 | 5298 | Intron | NP_001304976.1 | |||
NM_001318048.1 | 5298 | Intron | NP_001304977.1 | |||
NM_007356.2 | 5298 | Intron | NP_031382.2 | |||
XM_011515975.1 | 5298 | Silent Mutation | CAA,CAT | Q,H 1738 | XP_011514277.1 | |
XM_011515978.1 | 5298 | Intron | XP_011514280.1 | |||
XM_011515979.2 | 5298 | Intron | XP_011514281.1 | |||
XM_011515980.2 | 5298 | Intron | XP_011514282.1 | |||
XM_017011879.1 | 5298 | Silent Mutation | CAA,CAT | Q,H 1738 | XP_016867368.1 | |
XM_017011880.1 | 5298 | Intron | XP_016867369.1 |
Set Membership: |
HapMap |