Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL13 - methyltransferase like 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007239.1 | 43 | Intron | NP_001007240.1 | |||
NM_014955.2 | 43 | Intron | NP_055770.1 | |||
NM_015935.4 | 43 | Intron | NP_057019.3 | |||
XM_017001455.1 | 43 | Intron | XP_016856944.1 | |||
XM_017001456.1 | 43 | Intron | XP_016856945.1 | |||
XM_017001457.1 | 43 | Intron | XP_016856946.1 | |||
XM_017001458.1 | 43 | UTR 5 | XP_016856947.1 | |||
XM_017001459.1 | 43 | Intron | XP_016856948.1 |