Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606953 MIM: 613898 MIM: 616143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALE - UDP-galactose-4-epimerase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMGCL - 3-hydroxymethyl-3-methylglutaryl-CoA lyase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYPLA2 - lysophospholipase II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007260.2 | 548 | Intron | NP_009191.1 | |||
XM_005245728.4 | 548 | UTR 5 | XP_005245785.1 | |||
XM_017000165.1 | 548 | Intron | XP_016855654.1 |
PITHD1 - PITH domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |