Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAAGTGGTGACACACGGTTACT[T/G]CGGCTTGGGTGTCGGTGACCAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372493 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC105372493 - uncharacterized LOC105372493 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMF1 - proteasome inhibitor subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323407.1 | 288 | Intron | NP_001310336.1 | |||
NM_001323408.1 | 288 | Missense Mutation | TGC,TTC | C,F 36 | NP_001310337.1 | |
NM_001323409.1 | 288 | Missense Mutation | TGC,TTC | C,F 36 | NP_001310338.1 | |
NM_001323410.1 | 288 | Missense Mutation | TGC,TTC | C,F 36 | NP_001310339.1 | |
NM_006814.4 | 288 | Missense Mutation | TGC,TTC | C,F 36 | NP_006805.2 | |
NM_178578.3 | 288 | Missense Mutation | TGC,TTC | C,F 36 | NP_848693.2 |
Set Membership: |
HapMap Validated |