Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTGTTGCGGTCACAGGCATGAC[T/C]TAGGGTAGGATGGTGCTGCCCGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BZW1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BZW1 - basic leucine zipper and W2 domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207067.1 | Intron | NP_001193996.1 | ||||
NM_001207069.1 | Intron | NP_001193998.1 | ||||
NM_001321688.1 | Intron | NP_001308617.1 | ||||
NM_001321690.1 | Intron | NP_001308619.1 | ||||
NM_001321691.1 | Intron | NP_001308620.1 | ||||
NM_001321693.1 | Intron | NP_001308622.1 | ||||
NM_001321694.1 | Intron | NP_001308623.1 | ||||
NM_014670.3 | Intron | NP_055485.2 | ||||
XM_017005393.1 | Intron | XP_016860882.1 |
LOC101927795 - uncharacterized LOC101927795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |