Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137570 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ42351 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLJ42351 - uncharacterized LOC400999 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC20A1 - solute carrier family 20 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005415.4 | 1117 | Intron | NP_005406.3 | |||
XM_017004768.1 | 1117 | Intron | XP_016860257.1 | |||
XM_017004769.1 | 1117 | UTR 5 | XP_016860258.1 | |||
XM_017004770.1 | 1117 | Intron | XP_016860259.1 | |||
XM_017004771.1 | 1117 | Intron | XP_016860260.1 |
Set Membership: |
HapMap |