Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602989 MIM: 606913 | ||||||||||||||||||||
Literature Links: |
CLK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLK2 - CDC like kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001294338.1 | Intron | NP_001281267.1 | ||||
NM_001294339.1 | Intron | NP_001281268.1 | ||||
NM_003993.3 | Intron | NP_003984.2 | ||||
XM_005244878.1 | Intron | XP_005244935.1 | ||||
XM_011509143.1 | Intron | XP_011507445.1 | ||||
XM_011509144.1 | Intron | XP_011507446.1 | ||||
XM_011509147.2 | Intron | XP_011507449.1 | ||||
XM_017000241.1 | Intron | XP_016855730.1 | ||||
XM_017000242.1 | Intron | XP_016855731.1 | ||||
XM_017000243.1 | Intron | XP_016855732.1 | ||||
XM_017000244.1 | Intron | XP_016855733.1 |
FAM189B - family with sequence similarity 189 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCAMP3 - secretory carrier membrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005698.3 | Intron | NP_005689.2 | ||||
NM_052837.2 | Intron | NP_443069.1 | ||||
XM_006711105.3 | Intron | XP_006711168.1 | ||||
XM_006711106.3 | Intron | XP_006711169.1 | ||||
XM_016999991.1 | Intron | XP_016855480.1 |