Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608003 MIM: 610088 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIPK1 - homeodomain interacting protein kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OLFML3 - olfactomedin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286352.1 | 297 | UTR 5 | NP_001273281.1 | |||
NM_001286353.1 | 297 | Intron | NP_001273282.1 | |||
NM_020190.3 | 297 | Intron | NP_064575.1 | |||
XM_017001848.1 | 297 | UTR 5 | XP_016857337.1 |
Set Membership: |
HapMap JSNP |