Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
|||||||||||||||||||||||
Phenotype: |
MIM: 191311 | |||||||||||||||||||||||
Literature Links: |
DDR2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
DDR2 - discoidin domain receptor tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014796.1 | 167 | UTR 5 | NP_001014796.1 | |||
NM_006182.2 | 167 | UTR 5 | NP_006173.2 | |||
XM_006711344.3 | 167 | Intron | XP_006711407.1 | |||
XM_011509586.2 | 167 | Intron | XP_011507888.1 | |||
XM_011509587.2 | 167 | Intron | XP_011507889.1 | |||
XM_011509588.2 | 167 | UTR 5 | XP_011507890.1 |
Set Membership: |
HapMap JSNP |