Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBM27 - RNA binding motif protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018989.1 | Intron | NP_061862.1 | ||||
XM_005268466.1 | Intron | XP_005268523.1 | ||||
XM_005268467.1 | Intron | XP_005268524.1 | ||||
XM_005268468.1 | Intron | XP_005268525.1 | ||||
XM_006714787.1 | Intron | XP_006714850.1 | ||||
XM_011537661.1 | Intron | XP_011535963.1 | ||||
XM_011537662.1 | Intron | XP_011535964.1 | ||||
XM_017009587.1 | Intron | XP_016865076.1 |
Set Membership: |
HapMap |