Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602281 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAPLN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HAPLN3 - hyaluronan and proteoglycan link protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFGE8 - milk fat globule-EGF factor 8 protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114614.2 | 1334 | UTR 3 | NP_001108086.1 | |||
NM_001310319.1 | 1334 | UTR 3 | NP_001297248.1 | |||
NM_001310320.1 | 1334 | UTR 3 | NP_001297249.1 | |||
NM_001310321.1 | 1334 | UTR 3 | NP_001297250.1 | |||
NM_005928.3 | 1334 | UTR 3 | NP_005919.2 | |||
XM_017022206.1 | 1334 | Intron | XP_016877695.1 |
Set Membership: |
HapMap |