Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCAGTGTTTCCCTCCGAGGGCA[C/G]TGCGTGCCGCGCAGGCGCAAAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605358 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2A1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2A1L - general transcription factor IIA subunit 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193487.2 | 50 | UTR 5 | NP_001180416.1 | |||
NM_006872.4 | 50 | UTR 5 | NP_006863.2 |
STON1-GTF2A1L - STON1-GTF2A1L readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198593.1 | 50 | Intron | NP_001185522.1 | |||
NM_001198594.1 | 50 | Intron | NP_001185523.1 | |||
NM_172311.2 | 50 | Intron | NP_758515.1 |