Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146735 MIM: 601311 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFBP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IGFBP6 - insulin like growth factor binding protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOAT2 - sterol O-acyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003578.3 | Intron | NP_003569.1 | ||||
XM_011538849.2 | Intron | XP_011537151.1 | ||||
XM_011538850.2 | Intron | XP_011537152.1 | ||||
XM_011538851.2 | Intron | XP_011537153.1 |
Set Membership: |
HapMap Validated |