Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142910 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGCR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HMGCR - 3-hydroxy-3-methylglutaryl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000859.2 | Intron | NP_000850.1 | ||||
NM_001130996.1 | Intron | NP_001124468.1 | ||||
XM_011543357.1 | Intron | XP_011541659.1 | ||||
XM_011543358.1 | Intron | XP_011541660.1 | ||||
XM_011543359.1 | Intron | XP_011541661.1 |
Set Membership: |
HapMap |