Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147045 MIM: 604947 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCAR PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FCAR - Fc fragment of IgA receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002000.3 | Intron | NP_001991.1 | ||||
NM_133269.3 | Intron | NP_579803.1 | ||||
NM_133271.3 | Intron | NP_579805.1 | ||||
NM_133272.3 | Intron | NP_579806.1 | ||||
NM_133273.3 | Intron | NP_579807.1 | ||||
NM_133274.3 | Intron | NP_579808.1 | ||||
NM_133277.3 | Intron | NP_579811.1 | ||||
NM_133278.3 | Intron | NP_579812.1 | ||||
XM_011526625.2 | Intron | XP_011524927.1 | ||||
XM_017026473.1 | Intron | XP_016881962.1 | ||||
XM_017026474.1 | Intron | XP_016881963.1 |
KIR3DL2 - killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |