Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609854 MIM: 613766 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPCDC PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPCDC - phosphopantothenoylcysteine decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301101.1 | Intron | NP_001288030.1 | ||||
NM_001301102.1 | Intron | NP_001288031.1 | ||||
NM_001301103.1 | Intron | NP_001288032.1 | ||||
NM_001301104.1 | Intron | NP_001288033.1 | ||||
NM_001301105.1 | Intron | NP_001288034.1 | ||||
NM_021823.4 | Intron | NP_068595.3 | ||||
XM_017022461.1 | Intron | XP_016877950.1 | ||||
XM_017022462.1 | Intron | XP_016877951.1 | ||||
XM_017022463.1 | Intron | XP_016877952.1 | ||||
XM_017022464.1 | Intron | XP_016877953.1 | ||||
XM_017022465.1 | Intron | XP_016877954.1 |
SCAMP5 - secretory carrier membrane protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |