Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604071 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGKH - diacylglycerol kinase eta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204504.2 | Intron | NP_001191433.1 | ||||
NM_001204505.2 | Intron | NP_001191434.1 | ||||
NM_001204506.2 | Intron | NP_001191435.1 | ||||
NM_001297429.1 | Intron | NP_001284358.1 | ||||
NM_152910.5 | Intron | NP_690874.2 | ||||
NM_178009.4 | Intron | NP_821077.1 |