Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGGAGTGTGAAGAGAGCTTCAGA[C/T]GTCAAACATTTCCTAGAACCTGGTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600558 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373805 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105373805 - uncharacterized LOC105373805 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT4 - signal transducer and activator of transcription 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243835.1 | 2686 | UTR 3 | NP_001230764.1 | |||
NM_003151.3 | 2686 | UTR 3 | NP_003142.1 | |||
XM_006712719.3 | 2686 | UTR 3 | XP_006712782.1 | |||
XM_011511705.2 | 2686 | UTR 3 | XP_011510007.1 | |||
XM_017004784.1 | 2686 | Intron | XP_016860273.1 |
Set Membership: |
HapMap |