Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103390 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHNAK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AHNAK - AHNAK nucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001620.2 | 975 | Intron | NP_001611.1 | |||
NM_024060.3 | 975 | UTR 3 | NP_076965.2 | |||
XM_005274240.1 | 975 | Intron | XP_005274297.1 | |||
XM_005274241.2 | 975 | Intron | XP_005274298.1 | |||
XM_005274245.1 | 975 | Intron | XP_005274302.1 | |||
XM_017018270.1 | 975 | Intron | XP_016873759.1 | |||
XM_017018271.1 | 975 | Intron | XP_016873760.1 | |||
XM_017018272.1 | 975 | Intron | XP_016873761.1 | |||
XM_017018273.1 | 975 | Intron | XP_016873762.1 |