Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176875 MIM: 603761 MIM: 610831 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1CA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP1CA - protein phosphatase 1 catalytic subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAD9A - RAD9 checkpoint clamp component A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D10C - TBC1 domain family member 10C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256508.1 | 379 | Silent Mutation | TCC,TCT | S,S 21 | NP_001243437.1 | |
NM_198517.3 | 379 | Silent Mutation | TCC,TCT | S,S 21 | NP_940919.1 | |
XM_006718538.3 | 379 | Silent Mutation | TCC,TCT | S,S 21 | XP_006718601.1 | |
XM_006718539.3 | 379 | Silent Mutation | TCC,TCT | S,S 21 | XP_006718602.1 | |
XM_006718540.3 | 379 | Silent Mutation | TCC,TCT | S,S 21 | XP_006718603.1 | |
XM_006718541.3 | 379 | UTR 5 | XP_006718604.1 | |||
XM_006718542.3 | 379 | UTR 5 | XP_006718605.1 | |||
XM_006718543.2 | 379 | UTR 5 | XP_006718606.1 | |||
XM_006718544.3 | 379 | UTR 5 | XP_006718607.1 | |||
XM_011545001.2 | 379 | UTR 5 | XP_011543303.1 | |||
XM_011545002.2 | 379 | Silent Mutation | TCC,TCT | S,S 21 | XP_011543304.1 |
Set Membership: |
HapMap |