Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGGCTTGGGAAACATGGGGAT[C/T]GCTGTGTGTACGATGTTCATTGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607226 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HDAC11 - histone deacetylase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136041.2 | Intron | NP_001129513.1 | ||||
NM_024827.3 | Intron | NP_079103.2 | ||||
XM_005265491.1 | Intron | XP_005265548.1 | ||||
XM_005265492.1 | Intron | XP_005265549.1 | ||||
XM_006713338.1 | Intron | XP_006713401.1 | ||||
XM_011534131.1 | Intron | XP_011532433.1 | ||||
XM_011534132.2 | Intron | XP_011532434.2 | ||||
XM_011534135.1 | Intron | XP_011532437.1 | ||||
XM_017007234.1 | Intron | XP_016862723.1 |
HDAC11-AS1 - HDAC11 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |