Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608660 | ||||||||||||||||||||
Literature Links: |
INSIG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INSIG2 - insulin induced gene 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321329.1 | 72 | Intron | NP_001308258.1 | |||
NM_001321330.1 | 72 | UTR 5 | NP_001308259.1 | |||
NM_001321331.1 | 72 | UTR 5 | NP_001308260.1 | |||
NM_001321332.1 | 72 | Intron | NP_001308261.1 | |||
NM_001321333.1 | 72 | UTR 5 | NP_001308262.1 | |||
NM_016133.3 | 72 | UTR 5 | NP_057217.2 |