Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTAGCTGAATGAAGAAATTACT[C/G]AACAGTGTAGTGACAGGTCAGATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611817 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLRC4-KLRK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLRC4-KLRK1 - KLRC4-KLRK1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199805.1 | Intron | NP_001186734.1 |
KLRK1 - killer cell lectin like receptor K1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007360.3 | Intron | NP_031386.2 |
LOC101928100 - uncharacterized LOC101928100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |