Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609477 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMCO4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SMCO4 - single-pass membrane protein with coiled-coil domains 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020179.2 | Intron | NP_064564.1 | ||||
XM_011542907.1 | Intron | XP_011541209.1 | ||||
XM_011542908.2 | Intron | XP_011541210.1 | ||||
XM_011542909.1 | Intron | XP_011541211.1 | ||||
XM_011542910.2 | Intron | XP_011541212.1 | ||||
XM_011542911.2 | Intron | XP_011541213.1 | ||||
XM_017018019.1 | Intron | XP_016873508.1 | ||||
XM_017018020.1 | Intron | XP_016873509.1 |