Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612283 | ||||||||||||||||||||
Literature Links: |
MIR4783 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4783 - microRNA 4783 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PROC - protein C, inactivator of coagulation factors Va and VIIIa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000312.3 | Intron | NP_000303.1 | ||||
XM_005263717.4 | Intron | XP_005263774.1 | ||||
XM_017004505.1 | Intron | XP_016859994.1 | ||||
XM_017004506.1 | Intron | XP_016859995.1 |