Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611542 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARSB - arylsulfatase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000046.3 | 2161 | UTR 3 | NP_000037.2 | |||
NM_198709.2 | 2161 | Intron | NP_942002.1 | |||
XM_011543390.1 | 2161 | UTR 3 | XP_011541692.1 | |||
XM_011543391.2 | 2161 | Intron | XP_011541693.1 | |||
XM_011543392.2 | 2161 | Intron | XP_011541694.1 | |||
XM_011543393.1 | 2161 | Intron | XP_011541695.1 | |||
XM_017009471.1 | 2161 | Intron | XP_016864960.1 |