Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608034 MIM: 607066 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASPA - aspartoacylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA22 - spermatogenesis associated 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170695.1 | 4536 | Intron | NP_001164166.1 | |||
NM_001170696.1 | 4536 | Intron | NP_001164167.1 | |||
NM_001170697.1 | 4536 | Intron | NP_001164168.1 | |||
NM_001170698.1 | 4536 | Intron | NP_001164169.1 | |||
NM_001170699.1 | 4536 | Intron | NP_001164170.1 | |||
NM_001321336.1 | 4536 | Intron | NP_001308265.1 | |||
NM_001321337.1 | 4536 | Intron | NP_001308266.1 | |||
NM_032598.4 | 4536 | Intron | NP_115987.2 |
TRPV3 - transient receptor potential cation channel subfamily V member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258205.1 | 4536 | UTR 3 | NP_001245134.1 | |||
NM_145068.3 | 4536 | UTR 3 | NP_659505.1 |
Set Membership: |
HapMap |