Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300806 | ||||||||||||||||||||
Literature Links: |
AFF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AFF2 - AF4/FMR2 family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169122.1 | Intron | NP_001162593.1 | ||||
NM_001169123.1 | Intron | NP_001162594.1 | ||||
NM_001169124.1 | Intron | NP_001162595.1 | ||||
NM_001169125.1 | Intron | NP_001162596.1 | ||||
NM_001170628.1 | Intron | NP_001164099.1 | ||||
NM_002025.3 | Intron | NP_002016.2 |