Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604325 | ||||||||||||||||||||
Literature Links: |
PPP2R2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP2R2B - protein phosphatase 2 regulatory subunit Bbeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271899.1 | Intron | NP_001258828.1 | ||||
NM_001271900.1 | Intron | NP_001258829.1 | ||||
NM_001271948.1 | Intron | NP_001258877.1 | ||||
NM_181674.2 | Intron | NP_858060.2 | ||||
NM_181675.3 | Intron | NP_858061.2 | ||||
NM_181676.2 | Intron | NP_858062.1 | ||||
NM_181677.2 | Intron | NP_858063.1 | ||||
NM_181678.2 | Intron | NP_858064.1 |