Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCTGGCTGTCAGAAAGCAGGAC[C/T]GAGACTCGGTAGGATATAAGTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609101 MIM: 608048 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FBXO30 - F-box protein 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032145.4 | 5549 | Intron | NP_115521.3 | |||
XM_005267159.3 | 5549 | Intron | XP_005267216.1 | |||
XM_011536177.2 | 5549 | Intron | XP_011534479.1 | |||
XM_017011353.1 | 5549 | Intron | XP_016866842.1 |
LOC100507557 - uncharacterized LOC100507557 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHPRH - SNF2 histone linker PHD RING helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042683.2 | 5549 | Intron | NP_001036148.2 | |||
NM_173082.3 | 5549 | Intron | NP_775105.1 | |||
XM_006715439.3 | 5549 | UTR 3 | XP_006715502.1 | |||
XM_006715441.3 | 5549 | Intron | XP_006715504.1 | |||
XM_006715443.3 | 5549 | Intron | XP_006715506.1 | |||
XM_011535719.2 | 5549 | Intron | XP_011534021.1 | |||
XM_011535722.2 | 5549 | Intron | XP_011534024.1 | |||
XM_017010691.1 | 5549 | Intron | XP_016866180.1 | |||
XM_017010692.1 | 5549 | Intron | XP_016866181.1 | |||
XM_017010693.1 | 5549 | Intron | XP_016866182.1 | |||
XM_017010694.1 | 5549 | Intron | XP_016866183.1 | |||
XM_017010695.1 | 5549 | Intron | XP_016866184.1 | |||
XM_017010696.1 | 5549 | Intron | XP_016866185.1 |
Set Membership: |
HapMap Validated |