Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTATTTTCTCTTCACCATCAATG[C/T]CACTGCCACCTGTCTCCTACCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153440 MIM: 601022 MIM: 191160 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100287329 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100287329 - uncharacterized LOC100287329 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTA - lymphotoxin alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000595.3 | Intron | NP_000586.2 | ||||
NM_001159740.2 | Intron | NP_001153212.1 | ||||
XM_011514615.2 | Intron | XP_011512917.1 | ||||
XM_011514616.2 | Intron | XP_011512918.1 | ||||
XM_011514617.2 | Intron | XP_011512919.1 | ||||
XM_011514618.1 | Intron | XP_011512920.1 |
NFKBIL1 - NFKB inhibitor like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNF - tumor necrosis factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |