Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605502 MIM: 154365 MIM: 604943 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJC2 - DnaJ heat shock protein family (Hsp40) member C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129887.1 | 13 | UTR 5 | NP_001123359.1 | |||
NM_014377.1 | 13 | UTR 5 | NP_055192.1 | |||
XM_005250269.2 | 13 | UTR 5 | XP_005250326.1 | |||
XM_011516028.2 | 13 | UTR 5 | XP_011514330.1 | |||
XM_011516029.2 | 13 | UTR 5 | XP_011514331.1 | |||
XM_011516030.2 | 13 | Intron | XP_011514332.1 | |||
XM_017011968.1 | 13 | UTR 5 | XP_016867457.1 |
PSMC2 - proteasome 26S subunit, ATPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204453.1 | 13 | Intron | NP_001191382.1 | |||
NM_002803.3 | 13 | Intron | NP_002794.1 | |||
XM_005250505.1 | 13 | Intron | XP_005250562.1 |
SLC26A5 - solute carrier family 26 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |