Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600502 MIM: 611834 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGHMBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IGHMBP2 - immunoglobulin mu binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002180.2 | 325 | Missense Mutation | ACG,GCG | T,A 75 | NP_002171.2 | |
XM_005273974.2 | 325 | UTR 5 | XP_005274031.1 | |||
XM_005273975.3 | 325 | Intron | XP_005274032.1 | |||
XM_005273976.1 | 325 | Missense Mutation | ACG,GCG | T,A 75 | XP_005274033.1 | |
XM_011544994.1 | 325 | Intron | XP_011543296.1 | |||
XM_017017669.1 | 325 | UTR 5 | XP_016873158.1 | |||
XM_017017670.1 | 325 | Intron | XP_016873159.1 | |||
XM_017017671.1 | 325 | Missense Mutation | ACG,GCG | T,A 75 | XP_016873160.1 |
MRPL21 - mitochondrial ribosomal protein L21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |