Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608222 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADSL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADSL - adenylosuccinate lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000026.3 | 148 | Silent Mutation | CTG,TTG | L,L 42 | NP_000017.1 | |
NM_001123378.2 | 148 | Silent Mutation | CTG,TTG | L,L 42 | NP_001116850.1 | |
NM_001317923.1 | 148 | Intron | NP_001304852.1 | |||
XM_011529977.2 | 148 | Silent Mutation | CTG,TTG | L,L 42 | XP_011528279.1 | |
XM_011529979.1 | 148 | Silent Mutation | CTG,TTG | L,L 42 | XP_011528281.1 | |
XM_011529980.2 | 148 | Silent Mutation | CTG,TTG | L,L 42 | XP_011528282.1 | |
XM_017028636.1 | 148 | Silent Mutation | CTG,TTG | L,L 42 | XP_016884125.1 | |
XM_017028637.1 | 148 | Silent Mutation | CTG,TTG | L,L 42 | XP_016884126.1 | |
XM_017028638.1 | 148 | UTR 5 | XP_016884127.1 | |||
XM_017028639.1 | 148 | UTR 5 | XP_016884128.1 | |||
XM_017028640.1 | 148 | Intron | XP_016884129.1 |