Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606225 MIM: 165270 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAS1R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TAS1R1 - taste 1 receptor member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138697.3 | 141 | Intron | NP_619642.2 | |||
NM_177540.2 | 141 | Intron | NP_803884.1 | |||
XM_011542203.2 | 141 | Intron | XP_011540505.1 | |||
XM_011542206.2 | 141 | Intron | XP_011540508.1 | |||
XM_017002402.1 | 141 | Intron | XP_016857891.1 | |||
XM_017002403.1 | 141 | Intron | XP_016857892.1 |
ZBTB48 - zinc finger and BTB domain containing 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278647.1 | 141 | UTR 5 | NP_001265576.1 | |||
NM_001278648.1 | 141 | UTR 5 | NP_001265577.1 | |||
NM_005341.3 | 141 | UTR 5 | NP_005332.1 | |||
XM_017001110.1 | 141 | UTR 5 | XP_016856599.1 | |||
XM_017001111.1 | 141 | UTR 5 | XP_016856600.1 | |||
XM_017001112.1 | 141 | UTR 5 | XP_016856601.1 | |||
XM_017001113.1 | 141 | UTR 5 | XP_016856602.1 | |||
XM_017001114.1 | 141 | UTR 5 | XP_016856603.1 |
Set Membership: |
HapMap |