Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612491 MIM: 608769 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APIP - APAF1 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015957.3 | 589 | Intron | NP_057041.2 | |||
XM_011520154.2 | 589 | Intron | XP_011518456.1 | |||
XM_017017875.1 | 589 | Intron | XP_016873364.1 |
PDHX - pyruvate dehydrogenase complex component X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135024.1 | 589 | Intron | NP_001128496.1 | |||
NM_001166158.1 | 589 | Silent Mutation | CCC,CCT | P,P 21 | NP_001159630.1 | |
NM_003477.2 | 589 | Silent Mutation | CCC,CCT | P,P 21 | NP_003468.2 | |
XM_011520390.1 | 589 | Intron | XP_011518692.1 |