Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF641 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF641 - zinc finger protein 641 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172681.1 | 3442 | UTR 3 | NP_001166152.1 | |||
NM_001172682.1 | 3442 | UTR 3 | NP_001166153.1 | |||
NM_152320.2 | 3442 | UTR 3 | NP_689533.2 | |||
XM_005268637.3 | 3442 | UTR 3 | XP_005268694.1 | |||
XM_005268638.3 | 3442 | UTR 3 | XP_005268695.1 | |||
XM_005268639.2 | 3442 | UTR 3 | XP_005268696.1 | |||
XM_005268640.4 | 3442 | Intron | XP_005268697.1 | |||
XM_011537897.1 | 3442 | UTR 3 | XP_011536199.1 | |||
XM_011537898.1 | 3442 | UTR 3 | XP_011536200.1 | |||
XM_017018797.1 | 3442 | UTR 3 | XP_016874286.1 | |||
XM_017018798.1 | 3442 | UTR 3 | XP_016874287.1 | |||
XM_017018799.1 | 3442 | Intron | XP_016874288.1 |