Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAATCTTGTGACTATTGGGAAG[G/T]AGAGCAGCCAGAGGCTGAGGGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606720 MIM: 606057 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENDOU PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ENDOU - endonuclease, poly(U) specific | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105369748 - uncharacterized LOC105369748 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAPGEF3 - Rap guanine nucleotide exchange factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098531.2 | 5932 | UTR 3 | NP_001092001.1 | |||
NM_001098532.2 | 5932 | UTR 3 | NP_001092002.1 | |||
NM_006105.5 | 5932 | UTR 3 | NP_006096.2 | |||
XM_005268571.3 | 5932 | Intron | XP_005268628.1 | |||
XM_011537752.1 | 5932 | Intron | XP_011536054.1 | |||
XM_011537755.1 | 5932 | Intron | XP_011536057.1 | |||
XM_011537758.2 | 5932 | Intron | XP_011536060.1 | |||
XM_017018688.1 | 5932 | Intron | XP_016874177.1 |
Set Membership: |
HapMap |