Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606585 MIM: 147790 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENAM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ENAM - enamelin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JCHAIN - joining chain of multimeric IgA and IgM | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144646.3 | Intron | NP_653247.1 | ||||
XM_006714211.2 | Intron | XP_006714274.1 | ||||
XM_006714212.2 | Intron | XP_006714275.1 | ||||
XM_011531925.1 | Intron | XP_011530227.1 | ||||
XM_011531926.1 | Intron | XP_011530228.1 |
Set Membership: |
HapMap |