Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF226 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF226 - zinc finger protein 226 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032372.1 | Intron | NP_001027544.1 | ||||
NM_001032373.1 | Intron | NP_001027545.1 | ||||
NM_001032374.1 | Intron | NP_001027546.1 | ||||
NM_001146220.2 | Intron | NP_001139692.1 | ||||
NM_001319088.1 | Intron | NP_001306017.1 | ||||
NM_001319089.1 | Intron | NP_001306018.1 | ||||
NM_001319090.1 | Intron | NP_001306019.1 | ||||
NM_015919.3 | Intron | NP_057003.2 | ||||
NM_016444.2 | Intron | NP_057528.2 | ||||
XM_005259227.2 | Intron | XP_005259284.1 | ||||
XM_006723367.3 | Intron | XP_006723430.1 | ||||
XM_006723368.3 | Intron | XP_006723431.1 | ||||
XM_006723369.2 | Intron | XP_006723432.1 | ||||
XM_017027262.1 | Intron | XP_016882751.1 | ||||
XM_017027263.1 | Intron | XP_016882752.1 | ||||
XM_017027264.1 | Intron | XP_016882753.1 | ||||
XM_017027265.1 | Intron | XP_016882754.1 |
Set Membership: |
HapMap |