Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116880 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CTSL - cathepsin L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257971.1 | Intron | NP_001244900.1 | ||||
NM_001257972.1 | Intron | NP_001244901.1 | ||||
NM_001257973.1 | Intron | NP_001244902.1 | ||||
NM_001912.4 | Intron | NP_001903.1 | ||||
NM_145918.2 | Intron | NP_666023.1 | ||||
XM_005251716.3 | Intron | XP_005251773.1 | ||||
XM_011518263.1 | Intron | XP_011516565.1 | ||||
XM_017014293.1 | Intron | XP_016869782.1 |
Set Membership: |
HapMap |