Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606387 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOP1MT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TOP1MT - topoisomerase (DNA) I, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258446.1 | 1603 | Missense Mutation | CGG,TGG | R,W 427 | NP_001245375.1 | |
NM_001258447.1 | 1603 | Missense Mutation | CGG,TGG | R,W 427 | NP_001245376.1 | |
NM_052963.2 | 1603 | Missense Mutation | CGG,TGG | R,W 525 | NP_443195.1 | |
XM_005250781.3 | 1603 | Missense Mutation | CGG,TGG | R,W 525 | XP_005250838.1 |
Set Membership: |
HapMap |