Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610473 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPGAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LPGAT1 - lysophosphatidylglycerol acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320808.1 | 7442 | UTR 3 | NP_001307737.1 | |||
NM_014873.2 | 7442 | UTR 3 | NP_055688.1 | |||
XM_005273362.3 | 7442 | Intron | XP_005273419.1 | |||
XM_011510228.2 | 7442 | UTR 3 | XP_011508530.1 | |||
XM_011510229.2 | 7442 | UTR 3 | XP_011508531.1 | |||
XM_017003004.1 | 7442 | UTR 3 | XP_016858493.1 |
Set Membership: |
HapMap JSNP |